Methyl 5-(methylamino)nicotinate 95%
Dodavatel: Apollo Scientific
OR921634-1G
OR921634-250MG
APOSOR921634-1GEA
17990
CZK
APOSOR921634-1G
APOSOR921634-250MG
Methyl 5-(methylamino)nicotinate 95%
Methyl 5-(methylamino)nicotinate
Vzorec:
C₈H₁₀N₂O₂ MW: 166,177 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18256217 CAS číslo: 91702-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...