Tisk…

4-Chlorothiophene-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR921628-1G OR921628-250MG
APOSOR921628-1GEA 4800 CZK
APOSOR921628-1G APOSOR921628-250MG
4-Chlorothiophene-2-carbaldehyde 95%
4-Chlorothiophene-2-carbaldehyde
Vzorec: C₅H₃ClOS
MW: 146.595 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110670
CAS číslo: 57500-51-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR