Tisk…

3-(Ethylamino)phenol 95%

Dodavatel: Apollo Scientific

OR921595-10G OR921595-1G OR921595-5G
APOSOR921595-10GEA 21490 CZK
APOSOR921595-10G APOSOR921595-1G APOSOR921595-5G
3-(Ethylamino)phenol 95%
3-(Ethylamino)phenol
Vzorec: C₈H₁₁NO
MW: 137,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059617
CAS číslo: 621-31-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR