Tisk…

5-Methoxy-2-(methylthio)pyrimidin-4(3H)-one 95%

Dodavatel: Apollo Scientific

OR921539-1G OR921539-5G
APOSOR921539-1GEA 1210 CZK
APOSOR921539-1G APOSOR921539-5G
5-Methoxy-2-(methylthio)pyrimidin-4(3H)-one 95%
5-Methoxy-2-(methylthio)pyrimidin-4(3H)-one
Vzorec: C₆H₈N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD26935425
CAS číslo: 1671-08-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR