2-Bromo-5-methoxypyrimidine 98%
Dodavatel: Apollo Scientific
OR921490-1G
OR921490-250MG
APOSOR921490-1GEA
4900
CZK
APOSOR921490-1G
APOSOR921490-250MG
2-Bromo-5-methoxypyrimidine 98%
2-Bromo-5-methoxypyrimidine
Vzorec:
C₅H₅BrN₂O Storage Temperature: Ambient |
MDL Number:
MFCD12025462 CAS číslo: 1209459-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...