Tisk…

Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride 95%

Dodavatel: Apollo Scientific

OR921329-1G OR921329-250MG
APOSOR921329-1GEA 8570 CZK
APOSOR921329-1G APOSOR921329-250MG
Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride 95%
Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride
Vzorec: C₇H₁₃NO₂·ClH
MW: 179,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20685906
CAS číslo: 109837-32-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR