(4-Bromothiazol-2-yl)methanol 96%
Dodavatel: Apollo Scientific
OR921317-1G
APOSOR921317-1GEA
7770
CZK
APOSOR921317-1G
(4-Bromothiazol-2-yl)methanol 96%
(4-Bromothiazol-2-yl)methanol
Vzorec:
C₄H₄BrNOS MW: 194,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09746337 CAS číslo: 204513-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...