Tisk…

(S)-1-Isopropylpyrrolidin-3-amine 95%

Dodavatel: Apollo Scientific

OR921307-1G OR921307-250MG
APOSOR921307-1GEA 23970 CZK
APOSOR921307-1G APOSOR921307-250MG
(S)-1-Isopropylpyrrolidin-3-amine 95%
(S)-1-Isopropylpyrrolidin-3-amine
Vzorec: C₇H₁₆N₂
MW: 128,215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111639
CAS číslo: 1149384-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR