Tisk…

2-Norbornaneacetic acid 95%

Dodavatel: Apollo Scientific

OR921286-25G OR921286-5G OR921286-100MG OR921286-250MG
APOSOR921286-25GEA 0 CZK
APOSOR921286-25G APOSOR921286-5G APOSOR921286-100MG APOSOR921286-250MG
2-Norbornaneacetic acid 95%
2-Norbornaneacetic acid
Vzorec: C₉H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00167582
CAS číslo: 1007-01-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR