Tisk…

3-Isopropyl-1-methyl-1H-pyrazol-5-amine 90%

Dodavatel: Apollo Scientific

OR921265-1G OR921265-25G OR921265-5G
APOSOR921265-1GEA 3540 CZK
APOSOR921265-1G APOSOR921265-25G APOSOR921265-5G
3-Isopropyl-1-methyl-1H-pyrazol-5-amine 90%
3-Isopropyl-1-methyl-1H-pyrazol-5-amine
Vzorec: C₇H₁₃N₃
MW: 139,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664139
CAS číslo: 3702-12-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR