3-Chloro-1-methylpyridin-4(1H)-one 95%
Dodavatel: Apollo Scientific
OR920954-1G
OR920954-5G
APOSOR920954-1GEA
5200
CZK
APOSOR920954-1G
APOSOR920954-5G
3-Chloro-1-methylpyridin-4(1H)-one 95%
3-Chloro-1-methylpyridin-4(1H)-one
Vzorec:
C₆H₆ClNO MW: 143,571 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20327964 CAS číslo: 857201-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...