Tisk…

(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: [5-(Aminomethyl)-1-methyl-1H-pyrazol-3-yl]benzene, 5-(Aminomethyl)-1-methyl-3-phenyl-1H-pyrazole

OR9209-1G OR9209-250MG
APOSOR9209-1GEA 12340 CZK
APOSOR9209-1G APOSOR9209-250MG
(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine 97%
(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine
Vzorec: C₁₁H₁₃N₃
MW: 187,241 g/mol
Bod tání: 47,5…49.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08271943
CAS číslo: 876728-39-1
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR