Tisk…

Ethyl 3-(1H-imidazol-2-yl)propanoate 95%

Dodavatel: Apollo Scientific

OR920777-1G OR920777-250MG
APOSOR920777-1GEA 19840 CZK
APOSOR920777-1G APOSOR920777-250MG
Ethyl 3-(1H-imidazol-2-yl)propanoate 95%
Ethyl 3-(1H-imidazol-2-yl)propanoate
Vzorec: C₈H₁₂N₂O₂
MW: 168,193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20661662
CAS číslo: 172499-76-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR