5,6-Dichloro-2-methylpyridazin-3(2H)-one 95%
Dodavatel: Apollo Scientific
OR920667-1G
OR920667-250MG
APOSOR920667-1GEA
8770
CZK
APOSOR920667-1G
APOSOR920667-250MG
5,6-Dichloro-2-methylpyridazin-3(2H)-one 95%
5,6-Dichloro-2-methylpyridazin-3(2H)-one
Vzorec:
C₅H₄Cl₂N₂O MW: 179,004 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19443343 CAS číslo: 6794-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...