1-(1,1-Dimethoxy-ethyl)-2-methoxy-benzene 95%
Dodavatel: Apollo Scientific
OR920626-2G
OR920626-5G
APOSOR920626-2GEA
12400
CZK
APOSOR920626-2G
APOSOR920626-5G
1-(1,1-Dimethoxy-ethyl)-2-methoxy-benzene 95%
1-(1,1-Dimethoxy-ethyl)-2-methoxy-benzene
Vzorec:
C₁₁H₁₆O₃ MW: 196,243 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26131670 CAS číslo: 72244-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...