5-Bromo-1,2,4-triazole-3-carbonitrile 95%
Dodavatel: Apollo Scientific
OR920522-1G
OR920522-250MG
APOSOR920522-1GEA
4570
CZK
APOSOR920522-1G
APOSOR920522-250MG
5-Bromo-1,2,4-triazole-3-carbonitrile 95%
5-Bromo-1,2,4-triazole-3-carbonitrile
Vzorec:
C₃HBrN₄ Storage Temperature: Ambient |
MDL Number:
MFCD23381102 CAS číslo: 1427475-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...