Tisk…

1-Methyl-2-(methylsulphanyl)-1H-imidazole 95%

Dodavatel: Apollo Scientific

Synonyms: 1-Methyl-2-(methylthio)imidazole

OR920436-1G OR920436-5G
APOSOR920436-1GEA 1330 CZK
APOSOR920436-1G APOSOR920436-5G
1-Methyl-2-(methylsulphanyl)-1H-imidazole 95%
1-Methyl-2-(methylsulphanyl)-1H-imidazole
Vzorec: C₅H₈N₂S
MW: 128,195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25959069
CAS číslo: 14486-52-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR