Tisk…

3-Sec-butoxybenzaldehyde 95%

Dodavatel: Apollo Scientific

OR920300-1G OR920300-5G
APOSOR920300-1GEA 4770 CZK
APOSOR920300-1G APOSOR920300-5G
3-Sec-butoxybenzaldehyde 95%
3-Sec-butoxybenzaldehyde
Vzorec: C₁₁H₁₄O₂
MW: 178,228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08691916
CAS číslo: 915924-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR