N-(2-Acetyl-phenyl)-acetamide 98%
Dodavatel: Apollo Scientific
Synonyms:
N-(2-Acetylphenyl)acetamide
OR920016-1G
OR920016-5G
APOSOR920016-1GEA
1330
CZK
APOSOR920016-1G
APOSOR920016-5G
N-(2-Acetyl-phenyl)-acetamide 98%
N-(2-Acetyl-phenyl)-acetamide
Vzorec:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00032277 CAS číslo: 5234-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...