Methyl 2-(chloromethyl)acrylate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl2-(chloromethyl)acrylate
OR919913-1G
OR919913-5G
APOSOR919913-1GEA
4730
CZK
APOSOR919913-1G
APOSOR919913-5G
Methyl 2-(chloromethyl)acrylate 95%
Methyl 2-(chloromethyl)acrylate
Vzorec:
C₅H₇ClO₂ MW: 134.561 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11617809 CAS číslo: 922-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...