3-Methylbenzoyl cyanide 95%
Dodavatel: Apollo Scientific
OR919252-1G
OR919252-250MG
APOSOR919252-1GEA
14290
CZK
APOSOR919252-1G
APOSOR919252-250MG
3-Methylbenzoyl cyanide 95%
3-Methylbenzoyl cyanide
Vzorec:
C₉H₇NO MW: 145,158 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06658475 CAS číslo: 5955-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...