Tisk…

3-Methoxy-2-methyl-4H-pyran-4-one 95%

Dodavatel: Apollo Scientific

OR919247-5G
APOSOR919247-5GEA 7910 CZK
APOSOR919247-5G
3-Methoxy-2-methyl-4H-pyran-4-one 95%
3-Methoxy-2-methyl-4H-pyran-4-one
Vzorec: C₇H₈O₃
MW: 140,137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14553191
CAS číslo: 4780-14-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR