Tisk…
1-Methyl-prop-2-ynylamine hydrochloride 96%
Dodavatel: Apollo Scientific
Synonyms: But-3-yn-2-amine hydrochloride
OR919147-1G OR919147-250MG
APOSOR919147-1GEA 2150 CZK
APOSOR919147-1G APOSOR919147-250MG
1-Methyl-prop-2-ynylamine hydrochloride 96%
1-Methyl-prop-2-ynylamine hydrochloride
Vzorec: C₄H₇N·ClH
MW: 105,566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752577
CAS číslo: 42105-26-0