Tisk…

Ethyl 3-methyl-4-oxocrotonate 95%

Dodavatel: Apollo Scientific

OR919124-10G OR919124-1G OR919124-25G
APOSOR919124-10GEA 9330 CZK
APOSOR919124-10G APOSOR919124-1G APOSOR919124-25G
Ethyl 3-methyl-4-oxocrotonate 95%
Ethyl 3-methyl-4-oxocrotonate
Vzorec: C₇H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00043412
CAS číslo: 62054-49-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR