Tisk…

6-Hydroxy-2-benzoxazolinone 95%

Dodavatel: Apollo Scientific

OR919107-1G OR919107-5G
APOSOR919107-1GEA 5200 CZK
APOSOR919107-1G APOSOR919107-5G
6-Hydroxy-2-benzoxazolinone 95%
6-Hydroxy-2-benzoxazolinone
Vzorec: C₇H₅NO₃
MW: 151,119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369767
CAS číslo: 78213-03-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR