Tisk…
1-Methyl-1H-imidazol-4-amine hydrochloride 96%
Dodavatel: Apollo Scientific
OR919102-1G OR919102-250MG
APOSOR919102-1GEA 3710 CZK
APOSOR919102-1G APOSOR919102-250MG
1-Methyl-1H-imidazol-4-amine hydrochloride 96%
1-Methyl-1H-imidazol-4-amine hydrochloride
Vzorec: C₄H₇N₃·ClH
MW: 133,579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21340310
CAS číslo: 89088-69-7