2-Methyl-1-phenylpropan-1-amine hydrochloride 95%
Dodavatel: Apollo Scientific
OR919063-1G
OR919063-5G
APOSOR919063-1GEA
9330
CZK
APOSOR919063-1G
APOSOR919063-5G
2-Methyl-1-phenylpropan-1-amine hydrochloride 95%
2-Methyl-1-phenylpropan-1-amine hydrochloride
Vzorec:
C₁₀H₁₅N·ClH Storage Temperature: Ambient |
MDL Number:
MFCD18252291 CAS číslo: 24290-47-9 |