Tisk…

2-Bromo-[1,2,4]triazolo[1,5-a]pyridine 95%

Dodavatel: Apollo Scientific

OR919016-100MG OR919016-250MG
APOSOR919016-100MGEA 7250 CZK
APOSOR919016-100MG APOSOR919016-250MG
2-Bromo-[1,2,4]triazolo[1,5-a]pyridine 95%
2-Bromo-[1,2,4]triazolo[1,5-a]pyridine
Vzorec: C₆H₄BrN₃
Storage Temperature: Ambient
MDL Number: MFCD19441933
CAS číslo: 1021019-03-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR