5-Diethylamino-furan-2-carbaldehyde 95%
Dodavatel: Apollo Scientific
Synonyms:
5-(Diethylamino)furan-2-carbaldehyde
OR918523-1G
OR918523-250MG
APOSOR918523-1GEA
4200
CZK
APOSOR918523-1G
APOSOR918523-250MG
5-Diethylamino-furan-2-carbaldehyde 95%
5-Diethylamino-furan-2-carbaldehyde
Vzorec:
C₉H₁₃NO₂ MW: 167.205 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02614961 CAS číslo: 22868-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...