Methyl 1-sec-butyl-5-oxopyrrolidine-3-carboxylate 95%
Dodavatel: Apollo Scientific
OR918478-1G
OR918478-5G
APOSOR918478-1GEA
5200
CZK
APOSOR918478-1G
APOSOR918478-5G
Methyl 1-sec-butyl-5-oxopyrrolidine-3-carboxylate 95%
Methyl 1-sec-butyl-5-oxopyrrolidine-3-carboxylate
Vzorec:
C₁₀H₁₇NO₃ MW: 199.247 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21641342 CAS číslo: 1363166-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...