Tisk…

1-(2-Amino-5-chlorophenyl)ethanone 98%

Dodavatel: Apollo Scientific

OR918402-250MG OR918402-5G
APOSOR918402-250MGEA 4170 CZK
APOSOR918402-250MG APOSOR918402-5G
1-(2-Amino-5-chlorophenyl)ethanone 98%
1-(2-Amino-5-chlorophenyl)ethanone
Vzorec: C₈H₈ClNO
MW: 169,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07434611
CAS číslo: 1685-19-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR