Tisk…

1-(Pyridin-3-yl)cyclopropanecarboxylic acid 97%

Dodavatel: Apollo Scientific

OR918384-1G OR918384-250MG
APOSOR918384-1GEA 16340 CZK
APOSOR918384-1G APOSOR918384-250MG
1-(Pyridin-3-yl)cyclopropanecarboxylic acid 97%
1-(Pyridin-3-yl)cyclopropanecarboxylic acid
Vzorec: C₉H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD13188647
CAS číslo: 610791-39-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR