Tisk…

7-Methyl-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR918376-1G OR918376-250MG
APOSOR918376-1GEA 15310 CZK
APOSOR918376-1G APOSOR918376-250MG
7-Methyl-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde 95%
7-Methyl-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
Vzorec: C₁₀H₁₀O₃
MW: 178,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053948
CAS číslo: 724791-20-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR