4-tert-Butyl-1H-pyrazole 98%
Dodavatel: Apollo Scientific
Synonyms:
4-(tert-Butyl)-1H-pyrazole
OR918189-1G
APOSOR918189-1GEA
4300
CZK
APOSOR918189-1G
4-tert-Butyl-1H-pyrazole 98%
4-tert-Butyl-1H-pyrazole
Vzorec:
C₇H₁₂N₂ MW: 124,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847492 CAS číslo: 105285-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...