2-Methyl-6-nitro-1H-indole 95%
Dodavatel: Apollo Scientific
OR918153-1G
OR918153-5G
APOSOR918153-1GEA
2530
CZK
APOSOR918153-1G
APOSOR918153-5G
2-Methyl-6-nitro-1H-indole 95%
2-Methyl-6-nitro-1H-indole
Vzorec:
C₉H₈N₂O₂ MW: 176.172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13178656 CAS číslo: 3484-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...