Tisk…

1-(2-Propyn-1-yl)-1H-pyrrole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR918148-1G OR918148-250MG
APOSOR918148-1GEA 15310 CZK
APOSOR918148-1G APOSOR918148-250MG
1-(2-Propyn-1-yl)-1H-pyrrole-2-carbaldehyde 95%
1-(2-Propyn-1-yl)-1H-pyrrole-2-carbaldehyde
Vzorec: C₈H₇NO
Storage Temperature: Ambient
MDL Number: MFCD11695061
CAS číslo: 104501-02-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR