Tisk…

2-(5-Methylpyridin-2-yl)acetic acid 95%

Dodavatel: Apollo Scientific

OR918131-1G OR918131-250MG
APOSOR918131-1GEA 14680 CZK
APOSOR918131-1G APOSOR918131-250MG
2-(5-Methylpyridin-2-yl)acetic acid 95%
2-(5-Methylpyridin-2-yl)acetic acid
Vzorec: C₈H₉NO₂
MW: 151,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09924932
CAS číslo: 848093-05-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR