Tisk…

Ethyl 2-(methylsulphonyl)propanoate 95%

Dodavatel: Apollo Scientific

OR918027-1G OR918027-250MG
APOSOR918027-1GEA 19840 CZK
APOSOR918027-1G APOSOR918027-250MG
Ethyl 2-(methylsulphonyl)propanoate 95%
Ethyl 2-(methylsulphonyl)propanoate
Vzorec: C₆H₁₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD16045025
CAS číslo: 73017-82-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR