Tisk…

5-Hydroxy-1H-indole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR918002-1G OR918002-250MG
APOSOR918002-1GEA 6290 CZK
APOSOR918002-1G APOSOR918002-250MG
5-Hydroxy-1H-indole-3-carbaldehyde 95%
5-Hydroxy-1H-indole-3-carbaldehyde
Vzorec: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD06804528
CAS číslo: 3414-19-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR