4-Chloro-1-methyl-1H-indazole-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR917997-1G
OR917997-250MG
APOSOR917997-1GEA
15310
CZK
APOSOR917997-1G
APOSOR917997-250MG
4-Chloro-1-methyl-1H-indazole-3-carbaldehyde 95%
4-Chloro-1-methyl-1H-indazole-3-carbaldehyde
Vzorec:
C₉H₇ClN₂O MW: 194,618 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10686602 CAS číslo: 1033693-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...