Tisk…

2,7-Naphthyridine-4-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR917996-1G OR917996-250MG OR917996-5G OR917996-2G OR917996-10G
APOSOR917996-1GEA 0 CZK
APOSOR917996-1G APOSOR917996-250MG APOSOR917996-5G APOSOR917996-2G APOSOR917996-10G
2,7-Naphthyridine-4-carbaldehyde 95%
2,7-Naphthyridine-4-carbaldehyde
Vzorec: C₉H₆N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD15526648
CAS číslo: 10273-40-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR