Tisk…

Methyl 2-benzylacrylate 98%

Dodavatel: Apollo Scientific

OR917956-1G OR917956-5G
APOSOR917956-1GEA 2280 CZK
APOSOR917956-1G APOSOR917956-5G
Methyl 2-benzylacrylate 98%
Methyl 2-benzylacrylate
Vzorec: C₁₁H₁₂O₂
MW: 176,212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787445
CAS číslo: 3070-71-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR