Tisk…

5-Chloroisoindoline hydrochloride 95%

Dodavatel: Apollo Scientific

OR917946-1G OR917946-250MG
APOSOR917946-1GEA 2730 CZK
APOSOR917946-1G APOSOR917946-250MG
5-Chloroisoindoline hydrochloride 95%
5-Chloroisoindoline hydrochloride
Vzorec: C₈H₈ClN·ClH
MW: 190,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11223562
CAS číslo: 912999-79-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR