2,5-Dichlorobenzyl cyanide 98%
Dodavatel: Apollo Scientific
OR917907-25G
OR917907-100G
APOSOR917907-25GEA
1870
CZK
APOSOR917907-25G
APOSOR917907-100G
2,5-Dichlorobenzyl cyanide 98%
2,5-Dichlorobenzyl cyanide
Vzorec:
C₈H₅Cl₂N MW: 186,038 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02671209 CAS číslo: 3218-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...