Tisk…

7-Chloro-L-tryptophan 97%

Dodavatel: Apollo Scientific

OR917825-250MG OR917825-500MG
APOSOR917825-250MGEA 16770 CZK
APOSOR917825-250MG APOSOR917825-500MG
7-Chloro-L-tryptophan 97%
7-Chloro-L-tryptophan
Vzorec: C₁₁H₁₁ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09264322
CAS číslo: 73945-46-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR