(R)-2,4,6-Trimethylbenzenesulphinamide 97%
Dodavatel: Apollo Scientific
OR917824-1G
OR917824-250MG
APOSOR917824-1GEA
58850
CZK
APOSOR917824-1G
APOSOR917824-250MG
(R)-2,4,6-Trimethylbenzenesulphinamide 97%
(R)-2,4,6-Trimethylbenzenesulphinamide
Vzorec:
C₉H₁₃NOS Storage Temperature: Ambient |
MDL Number:
MFCD11707206 CAS číslo: 446021-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...