Tisk…

4-Methylbenzoyl cyanide 95%

Dodavatel: Apollo Scientific

OR917809-1G OR917809-250MG
APOSOR917809-1GEA 9330 CZK
APOSOR917809-1G APOSOR917809-250MG
4-Methylbenzoyl cyanide 95%
4-Methylbenzoyl cyanide
Vzorec: C₉H₇NO
MW: 145,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00182410
CAS číslo: 14271-73-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR