Ethyl 1H-1,2,3-triazole-4-carboxylate 95%
Dodavatel: Apollo Scientific
OR917782-1G
APOSOR917782-1GEA
2070
CZK
APOSOR917782-1G
Ethyl 1H-1,2,3-triazole-4-carboxylate 95%
Ethyl 1H-1,2,3-triazole-4-carboxylate
Vzorec:
C₅H₇N₃O₂ MW: 141,128 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08669518 CAS číslo: 40594-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...