Tisk…

3-(tert-Butyl)-4-hydroxybenzonitrile 96%

Dodavatel: Apollo Scientific

OR917655-1G OR917655-250MG
APOSOR917655-1GEA 4200 CZK
APOSOR917655-1G APOSOR917655-250MG
3-(tert-Butyl)-4-hydroxybenzonitrile 96%
3-(tert-Butyl)-4-hydroxybenzonitrile
Vzorec: C₁₁H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD16999179
CAS číslo: 4910-04-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR