Benzyl glycolate 97%
Dodavatel: Apollo Scientific
OR917631-100G
OR917631-25G
APOSOR917631-100GEA
2780
CZK
APOSOR917631-100G
APOSOR917631-25G
Benzyl glycolate 97%
Benzyl glycolate
Vzorec:
C₉H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00191776 CAS číslo: 30379-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...